missing translation for 'onlineSavingsMsg'
Learn More

Oleamide

Catalog No. p-7214773
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467650050 5 g
AC467650250 25 g
AC467651000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC467650050 Supplier Thermo Scientific Chemicals Supplier No. 467650050
Only null left

CAS: 301-02-0 | C18H35NO | 281.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 301-02-0
Molecular Formula C18H35NO
Molecular Weight (g/mol) 281.48
InChI Key FATBGEAMYMYZAF-MDZDMXLPSA-N
IUPAC Name (9E)-octadec-9-enamide
SMILES CCCCCCCC\C=C\CCCCCCCC(N)=O

Specifications

Melting Point 66.0°C to 76.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in ether, ethanol
Formula Weight 281.48
Chemical Name or Material Oleamide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 206-103-9

RUO – Research Use Only