missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octanophenone, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 279360250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Octanophenone | |
99.0 | |
0.93g/mL | |
99+% | |
C14H20O | |
1.5024 to 1.5054 (20°C, 589nm) | |
25g | |
UDEVCZRUNOLVLU-UHFFFAOYSA-N | |
1-phenyloctan-1-one | |
74291 | |
99+% |
1674-37-9 | |
100.0 | |
Authentic | |
Glass Bottle | |
C6H5CO(CH2)6CH3 | |
MFCD00003554 | |
octanophenone, heptyl phenyl ketone, 1-octanone, 1-phenyl, caprylophenone, n-octanophenone, ketone, heptyl phenyl, 4-n-butylbutyrophenone, 1-phenyl-octan-1-one, acmc-1axlq | |
CCCCCCCC(=O)C1=CC=CC=C1 | |
204.31 | |
204.31 |
Chemical Identifiers
1674-37-9 | |
204.31 | |
UDEVCZRUNOLVLU-UHFFFAOYSA-N | |
74291 | |
CCCCCCCC(=O)C1=CC=CC=C1 |
C14H20O | |
MFCD00003554 | |
octanophenone, heptyl phenyl ketone, 1-octanone, 1-phenyl, caprylophenone, n-octanophenone, ketone, heptyl phenyl, 4-n-butylbutyrophenone, 1-phenyl-octan-1-one, acmc-1axlq | |
1-phenyloctan-1-one |
RUO â Research Use Only