missing translation for 'onlineSavingsMsg'
Learn More

Octanophenone, 99+%, Thermo Scientific™

Chemical Identifiers

CAS 1674-37-9
Molecular Formula C14H20O
Molecular Weight (g/mol) 204.31
MDL Number MFCD00003554
InChI Key UDEVCZRUNOLVLU-UHFFFAOYSA-N
Synonym octanophenone, heptyl phenyl ketone, 1-octanone, 1-phenyl, caprylophenone, n-octanophenone, ketone, heptyl phenyl, 4-n-butylbutyrophenone, 1-phenyl-octan-1-one, acmc-1axlq
PubChem CID 74291
IUPAC Name 1-phenyloctan-1-one
SMILES CCCCCCCC(=O)C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC279360050
SDS View Documents
Thermo Scientific Chemicals
279360050
5g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC279360250
SDS View Documents
Thermo Scientific Chemicals
279360250
25g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC279361000
SDS View Documents
Thermo Scientific Chemicals
279361000
100g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1674-37-9
204.31
UDEVCZRUNOLVLU-UHFFFAOYSA-N
74291
CCCCCCCC(=O)C1=CC=CC=C1
C14H20O
MFCD00003554
octanophenone, heptyl phenyl ketone, 1-octanone, 1-phenyl, caprylophenone, n-octanophenone, ketone, heptyl phenyl, 4-n-butylbutyrophenone, 1-phenyl-octan-1-one, acmc-1axlq
1-phenyloctan-1-one

Specifications

1674-37-9
100.0
Authentic
Glass Bottle
C6H5CO(CH2)6CH3
MFCD00003554
octanophenone, heptyl phenyl ketone, 1-octanone, 1-phenyl, caprylophenone, n-octanophenone, ketone, heptyl phenyl, 4-n-butylbutyrophenone, 1-phenyl-octan-1-one, acmc-1axlq
CCCCCCCC(=O)C1=CC=CC=C1
204.31
204.31
Octanophenone
99.0
0.93g/mL
99+%
C14H20O
1.5024 to 1.5054 (20°C, 589nm)
5g
UDEVCZRUNOLVLU-UHFFFAOYSA-N
1-phenyloctan-1-one
74291
99+%
Videos
SDS
Documents

Documents

RUO – Research Use Only