missing translation for 'onlineSavingsMsg'
Learn More

Octadecyltrimethoxysilane, 90%, Tech.

CAS: 3069-42-9 | C21H46O3Si | 374.69 g/mol

Supplier:  Thermo Scientific Chemicals 313601000

 View more versions of this product

Catalog No. AC313601000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 221-339-2

Recommended Storage : Product may become turbid on storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only