missing translation for 'onlineSavingsMsg'
Learn More

Octadecyltrimethoxysilane, 90%, Tech.

CAS: 3069-42-9 | C21H46O3Si | 374.69 g/mol

$235.19 - $831.20

Chemical Identifiers

CAS 3069-42-9
Molecular Formula C21H46O3Si
Molecular Weight (g/mol) 374.69
MDL Number MFCD00043060
InChI Key SLYCYWCVSGPDFR-UHFFFAOYSA-N
Synonym octadecyltrimethoxysilane, trimethoxy octadecyl silane, n-octadecyltrimethoxysilane, stearyltrimethoxysilane, silane, trimethoxyoctadecyl, octadecyl-trimethoxysilane, acmc-209hi7, ksc491g4l, trimethoxy octadecyl silane, technical grade
PubChem CID 76486
IUPAC Name trimethoxy(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](OC)(OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC313600250
SDS View Documents
Thermo Scientific Chemicals
313600250
25 mL Glass bottle
Each for $235.19
Only null left
 
AC313601000
SDS View Documents
Thermo Scientific Chemicals
313601000
100 mL Glass bottle
Each for $831.20
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3069-42-9
374.69
SLYCYWCVSGPDFR-UHFFFAOYSA-N
76486
CCCCCCCCCCCCCCCCCC[Si](OC)(OC)OC
C21H46O3Si
MFCD00043060
octadecyltrimethoxysilane, trimethoxy octadecyl silane, n-octadecyltrimethoxysilane, stearyltrimethoxysilane, silane, trimethoxyoctadecyl, octadecyl-trimethoxysilane, acmc-209hi7, ksc491g4l, trimethoxy octadecyl silane, technical grade
trimethoxy(octadecyl)silane

Specifications

3069-42-9
100.0
Colorless to Yellow
170°C (0.1 mmHg)
Authentic
Glass bottle
1.4380 to 1.44
MFCD00043060
0.88
Solubility in water: insoluble (reacts)
CCCCCCCCCCCCCCCCCC[Si](OC)(OC)OC
374.69
374.69
Technical
Octadecyltrimethoxysilane
88.0
13°C to 17°C
0.8800g/mL
139°C
88% min. Total octadecyltrimethoxysilanes (GC)
C21H46O3Si
CH3(CH2)17Si(OCH3)3
25 mL
octadecyltrimethoxysilane, trimethoxy octadecyl silane, n-octadecyltrimethoxysilane, stearyltrimethoxysilane, silane, trimethoxyoctadecyl, octadecyl-trimethoxysilane, acmc-209hi7, ksc491g4l, trimethoxy octadecyl silane, technical grade
SLYCYWCVSGPDFR-UHFFFAOYSA-N
trimethoxy(octadecyl)silane
76486
90%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 221-339-2

Recommended Storage : Product may become turbid on storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only