missing translation for 'onlineSavingsMsg'
Learn More

O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5403506

 View more versions of this product

Catalog No. AAH5403506


Only null left
Explore available promotions

Description

Description

Coupling reagent for peptide synthesis and the formation of other amides. Reactant for synthesis of liposomal contrast agents for magnetic resonance imaging, synthesis of protein labeling molecules, synthesis of thiol-reactive Cy5 derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coupling reagent for peptide synthesis and the formation of other amides. Reactant for synthesis of liposomal contrast agents for magnetic resonance imaging, synthesis of protein labeling molecules, synthesis of thiol-reactive Cy5 derivatives.

Solubility
Soluble in acetonitrile 0.5 g/mL.

Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only