Learn More
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98+%, Thermo Scientific Chemicals
$185.36 - $185.36
Chemical Identifiers
| CAS | 265651-18-1 |
|---|---|
| Molecular Formula | C9H16F6N3O3P |
| Molecular Weight (g/mol) | 359.21 |
| MDL Number | MFCD01863753 |
| InChI Key | STWZCCVNXFLDDD-UHFFFAOYSA-N |
| Synonym | hstu, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, ambotzrl-1039, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate |
| PubChem CID | 16211151 |
| IUPAC Name | [dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;hexafluorophosphate |
| SMILES | F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5403506
|
Thermo Scientific Chemicals
H5403506 |
5 g |
Each for $185.36
|
|
|||||
Description
Coupling reagent for peptide synthesis and the formation of other amides. Reactant for synthesis of liposomal contrast agents for magnetic resonance imaging, synthesis of protein labeling molecules, synthesis of thiol-reactive Cy5 derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCoupling reagent for peptide synthesis and the formation of other amides. Reactant for synthesis of liposomal contrast agents for magnetic resonance imaging, synthesis of protein labeling molecules, synthesis of thiol-reactive Cy5 derivatives.
Solubility
Soluble in acetonitrile 0.5 g/mL.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Chemical Identifiers
| 265651-18-1 | |
| 359.21 | |
| STWZCCVNXFLDDD-UHFFFAOYSA-N | |
| 16211151 | |
| F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C |
| C9H16F6N3O3P | |
| MFCD01863753 | |
| hstu, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, ambotzrl-1039, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate | |
| [dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;hexafluorophosphate |
Specifications
| 265651-18-1 | |
| C9H16F6N3O3P | |
| 5 g | |
| hstu, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium hexafluorophosphate, hstu n,n,n',n'-tetramethyl-o-n-succinimidyl uronium hexafluorophosphate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium hexafluorophosphate, ambotzrl-1039, o-n-succinimidyl-1,1,3,3-tetramethyluronium hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronim hexafluorophosphate, 2-succinimido-1,1,3,3-tetramethyluronium hexafluorophosphate, n,n,n',n'-tetramethyl-o-n-succinimidyl uroniumhexafluorophosphate | |
| STWZCCVNXFLDDD-UHFFFAOYSA-N | |
| [dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;hexafluorophosphate | |
| 16211151 | |
| ≥98% |
| ∽220°C (decomposition) | |
| MFCD01863753 | |
| Moisture sensitive | |
| Soluble in acetonitrile 0.5g/mL. | |
| F[P-](F)(F)(F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C | |
| 359.21 | |
| 359.21 | |
| O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium hexafluorophosphate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only