Learn More
Nystatin, 10 mg/ml in methanol, sterile-filtered, Thermo Scientific Chemicals

$105.75 - $342.47
Chemical Identifiers
CAS | 1400-61-9 |
---|---|
Molecular Formula | C47H75NO17 |
Molecular Weight (g/mol) | 926.107 |
MDL Number | MFCD00036240 |
InChI Key | VQOXZBDYSJBXMA-AWCIUYGFSA-N |
PubChem CID | 133640190 |
IUPAC Name | (1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carbShow More |
SMILES | CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAJ67369XF
|
Thermo Scientific Chemicals
J67369XF |
1 mL |
Each for $105.75
|
|
||||||
AAJ673698EQ
|
Thermo Scientific Chemicals
J673698EQ |
5 x 1 mL |
Pack for $342.47
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1400-61-9 | |
926.107 | |
VQOXZBDYSJBXMA-AWCIUYGFSA-N | |
(1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carbShow More |
C47H75NO17 | |
MFCD00036240 | |
133640190 | |
CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)OShow More |
Specifications
Nystatin | |
1400-61-9 | |
1 mL | |
MFCD00036240 | |
VQOXZBDYSJBXMA-AWCIUYGFSA-N | |
(1S,3R,6Z,18S,19R,20S,21S,25S,27S,29S,32R,33S,35S,37S,38R)-3-[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy-19,25,27,29,32,33,35,37-octahydroxy-18,20,21-trimethyl-23-oxo-22,39-dioxabicyclo[33.3.1]nonatriaconta-4,6,8,10,14,16-hexaene-38-carb | |
133640190 |
Liquid | |
Plastic Bottle | |
C47H75NO17 | |
UN1230 | |
CC1C=CC=CCCC=CC=CC=CC=CC(CC2C(C(CC(O2)(CC(C(CCC(CC(CC(CC(=O)OC(C(C1O)C)C)O)O)O)O)O)O)O)C(=O)O)OC3C(C(C(C(O3)C)O)N)O | |
926.107 | |
Nystatin |
Safety and Handling
GHS H Statement
H225-H311-H331-H370
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H311+H331-H370
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL
EINECSNumber : 215-749-0
RTECSNumber : RF5950000
TSCA : No
storageNote1 : −30° to −10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only