missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Flat Gel Loading Tips DFS Item

For easy and accurate loading of samples

Supplier:  Invitrogen™ LC1002

Catalog No. LC1002


Only null left
Explore available promotions

Description

Description

Tips are RNase/DNase-free and non-autoclavable. Standard, round can be used for easy and accurate loading of samples onto 1.0mm and 1.mm gels. Loading volume is 0.5 to 200 ÎĽL.

Applications:

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Greener Choice Claims
  • Environmental benefits include:
    • Source Reduction
    • Recyclable
Specifications

Specifications

Store at room temperature.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Eppendorf
No
Room Temperature
200 ÎĽL
Flat Tips
Universal Fit
Flat Gel Loading Tip
200/ pkg.
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.