Learn More
Honeywell™ North™ Nitri-Knit™ Gloves
Supported 12 in. nitrile gloves with interlocked knit liner feature an attached 15 in. sleeve for full arm protection.
Supplier:  Honeywell™ NK803ES11
 View more versions of this product
Catalog No. 19-160-0140
Only null left
Explore available promotions
Description
Description
  • High nitrile content offers resistance to cuts, snags, abrasions and punctures
  • Ideal for petrochemical refining, degreasing operations, automotive assembly and chemical handling
  • Resist petrochemicals, fats, oils, many acids and solvents
  • Free of latex proteins which can cause allergic reactions
  • Will not swell, sag or weaken from fat absorption
  • Interlocked knit liner provides comfort
Specifications
Specifications
21 CFR 170-199, ASTM D3389-05, 29 CFR 1915.157
No
Yes
11
No
Elasticated
Interlock knit
NK803ES/11
27 in.
EN 420, EN 374
Blue
Nitrile
EN 388
Automotive assembly, painting, chemical handling, processing, degreasing, dip tank cleanup, fishing industry, food processing, general maintenance work, printing cleanup, refinery (general), Solvents (general)
Rough
Lined
Nitri-Knit™
68 cm
Product Suggestions
Product Suggestions