missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Non Metals, plasma standard solution, Specpure™, As, B, P, S, Se, Si, Te at 100μg/mL

Manufacturer:  Alfa Aesar™ 44267AP

 View more versions of this product

Catalog No. AA44267AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Non Metals plasma standard solution
MFCD00144997
Liquid
500mL
As, B, P, S, Se, Si, Te at 100μg/mL
UN3264
Specpure™
Matrix: 5% HCl/tr. F-
Documents