missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[NH2Me2][(RuCl((S)-xylbinap))2(mu-Cl)3], TCI America™
Supplier: TCI America N10201G
Specifications
[NH2Me2][(RuCl((S)-xylbinap))2(mu-Cl)3] | |
C106H104Cl5NP4Ru2 | |
1 g | |
KEZRTMOWSKGUPV-UHFFFAOYSA-J | |
[1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;N-methylmethanamine;ruthenium(2+);tetrachloride;hydrochloride | |
131675154 | |
Crystalline Powder |
944451-10-9 | |
MFCD09753025 | |
Dimethylammonium Dichlorotri(mu-chloro)bis[(S)-(-)-2,2′C-bis[di-(3,5-xylyl)phosphino]-1,1′C-binaphthyl]diruthenate(II) | |
CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CNC.Cl.[Cl-].[Cl-].[C | |
1895.29 | |
1895.29 |
Chemical Identifiers
944451-10-9 | |
1895.29 | |
KEZRTMOWSKGUPV-UHFFFAOYSA-J | |
131675154 | |
CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CNC.Cl.[Cl-].[Cl-].[CShow More |
C106H104Cl5NP4Ru2 | |
MFCD09753025 | |
Dimethylammonium Dichlorotri(mu-chloro)bis[(S)-(-)-2,2′C-bis[di-(3,5-xylyl)phosphino]-1,1′C-binaphthyl]diruthenate(II)Show More | |
[1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;N-methylmethanamine;ruthenium(2+);tetrachloride;hydrochlorideShow More |
Safety and Handling
TSCA : No