missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide, ACS Reagent (DMF), Reagents
N,N-Dimethylformamide, ACS Reagent (DMF), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C119620055E
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
N,N-Dimethylformamide, ACS Reagent (DMF) | |
68-12-2 | |
152°C | |
Steel Drum | |
0.944 | |
Dimethylformamide, DMF, N,N-Dimethylmethanamide | |
CN(C)C=O | |
73.10 |
-60.5°C | |
Colorless to Slightly Yellow | |
55 gal. | |
Liquid | |
C3H7NO | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
ACS |
Chemical Identifiers
68-12-2 | |
73.10 | |
Dimethylformamide, DMF, N,N-Dimethylmethanamide | |
CN(C)C=O |
C3H7NO | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide |