missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, ACS Reagent (DMF), Reagents

Catalog No. p-200021130
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
55 gal.
Packaging:
Glass Bottle
Poly Bottle
Steel Drum
Steel Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01338009 20 L Steel Pail
01338008 4 L Poly Bottle
01338007 4 L Glass Bottle
01338006 500 mL Glass Bottle
01338005 55 gal. Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01338009 Supplier Labchem Inc Supplier No. C119620020E6
Only null left

N,N-Dimethylformamide, ACS Reagent (DMF), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym Dimethylformamide, DMF, N,N-Dimethylmethanamide
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -60.5°C
Color Colorless to Slightly Yellow
Boiling Point 152°C
Quantity 20 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.944
Grade ACS
Chemical Name or Material N,N-Dimethylformamide, ACS Reagent (DMF)