missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mycobacterium tuberculosis MPT64 Polyclonal Antibody
Supplier: Thermo Fisher Scientific PA5117509
Specifications
Mycobacterium tuberculosis MPT64 | |
Polyclonal | |
Unconjugated | |
mpt64 | |
mpt64 | |
Recombinant Mycobacterium tuberculosis Immunogenic protein MPT64 protein (24-228aa). | |
100 μg | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Western Blot | |
2.6 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P9WIN9 | |
Rabbit | |
Protein G | |
RUO | |
885925 | |
-20°C or -80°C if preferred | |
Liquid |