missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega MultiTox-Fluorâ„¢ Multiplex Cytotoxicity Assay
The MultiTox-Fluor Multiplex Cytotoxicity Assay is a single-reagent-addition, homogeneous, fluorescent assay that measures the number of live and dead cells simultaneously in culture wells.
$484.88 - $2785.40
Specifications
Content And Storage | -30°C to -10°C |
---|---|
Detection Method | Homogeneous, Fluorescence |
For Use With (Equipment) | Microplate Reader |
Description
- A Fluorescent Assay That Measures Both Cell Viability and Cytotoxicity
- Assay normalizes data for cell number
- Reduces false-positive and false-negative results
Order Info
Each 10mL aliquot is sufficient for 100 assays at 100μL/assay in a 96-well format or 400 assays at 25μL/assay in a 384-well format
Specifications
-30°C to -10°C | |
Homogeneous, Fluorescence | |
Microplate Reader |
Patent pending.