missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigmaâ„¢ MultiScreenHTS 384-Well Filter Plates
Use in automated, high throughput applications, including biological assays and sample preparation. MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.
$1049.88 - $4770.17
Specifications
Format | Standard |
---|---|
Membrane | Glass Fiber, Polyester Mesh |
Array | 16 x 24 |
Color | Opaque, White Bottom |
Material | Barex |
Catalog Number | Mfr. No. | Pore Size | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | Quantity | Price | Quantity | |||||
MZFCN0W10
|
MilliporeSigmaâ„¢
MZFCN0W10 |
1.2 μm | 10/Pk. |
Pack of 10 Each for $1,049.88
|
|
|||||
MZFBN0W50
|
MilliporeSigmaâ„¢
MZFBN0W50 |
1 μm | 50/Pk. |
Pack of 50 Each for $4,701.41
|
|
|||||
MZFBN0W10
|
MilliporeSigmaâ„¢
MZFBN0W10 |
1 μm | 10/Pk. |
Pack of 10 Each for $1,073.44
|
|
|||||
MZFCN0W50
|
MilliporeSigmaâ„¢
MZFCN0W50 |
1.2 μm | 50/Pk. |
Pack of 50 Each for $4,770.17
|
|
|||||
Description
For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.- Rigid sidewalls allows full compatibility with automated/robotic systems
- Unique plate design and individually sealed filter discs prevent cross-talk between wells
- Opaque plastic lids prevents light transfer between wells during direct scintillation counting
- Barcoding available
- Meet ANSIâ„¢/SBS standards
Specifications
Standard | |
16 x 24 | |
Barex | |
Non-sterile | |
1.28 cm | |
1.44 cm |
Glass Fiber, Polyester Mesh | |
Opaque, White Bottom | |
384 | |
1.28 x 8.55 x 1.44 cm | |
8.55 cm |