missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Epigen ELISA Kit
ELISA
Supplier: Invitrogen™ EM27RB
Description
Mouse Epigen quantitates mouse Epigen in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse Epigen.
Epigen is an EGF-related polypeptide growth factor that signals through the ErbB receptor-1. Promotes the growth of epithelial cells. May stimulate the phosphorylation of EGFR and mitogen-activated protein kinases. It is produced in several tissues, including the testis, liver, and heart, as well as in certain tumor cells. Epigen is mitogenic for fibroblasts and epithelial cells. Human Epigen is initially synthesized as a glycosylated 14.7 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a mature soluble sequence.Specifications
2310069M11Rik; ALGV3072; EPG; EPGN; epigen; Epithelial mitogen; epithelial mitogen homolog; PRO9904; RGD1560084; UNQ3072/PRO9904 | |
6.2 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
EPGN | |
<10 | |
HRP | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |
6.2 to 1,500 pg/mL | |
HRP | |
Plasma, Serum, Supernatant | |
71920 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Plasma 50 ÎĽL, Serum 50 ÎĽL, Supernatant 50 ÎĽL | |
Epigen | |
4 hrs 45 mins |