Learn More
Morphix Technologies Color Comparators for ChromAir™ Passive Sampling Systems
Use with ChromAir badge to determine the exact dose of a specific contaminant present in the workplace.
Supplier: Morphix Technologies 384003
Description
Color scale identically matches the colors formed on a ChromAir badge when it is exposed to a contaminant.- Badge slides in comparator for comparison to color scale insert
- Exact exposure dose is shown in the exposure dose window
Specifications
| To determine the exact dose of a specific contaminant present in the workplace | |
| Mercury | |
| 0.3 to 40ppm.Hour | |
| Color comparator |