Learn More
Methyl 2-(hydroxymethyl)acrylate, 85%, stabilized with 0.1 wt.% MEHQ, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 435900010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Environmental benefits include:
- Renewable Energy
Specifications
Methyl 2-(hydroxymethyl)acrylate | |
15484-46-5,150-19-6 | |
71°C to 72°C (2.0 Torr) | |
Authentic | |
Glass Bottle | |
MFCD12545894 | |
1.129 | |
RFUCOAQWQVDBEU-UHFFFAOYSA-N | |
methyl 2-(hydroxymethyl)prop-2-enoate | |
0.1 wt% Monomethylether hydroquinone | |
116.12 |
Stabilized | |
1.1290g/mL | |
90°C | |
84% min. (GC) | |
C5H8O3 | |
1g | |
methyl 2-hydroxymethyl acrylate, methyl 2-hydroxymethyl prop-2-enoate, methyl2-hydroxymethyl acrylate, methyl-2-hydroxymethyl-acrylate, pubchem17373, methyl 2-hydroxymethylacrylate, methyl alpha-hydroxymethylacrylate, 2-hydroxymethylacrylate methyl ester, 2-methylolacrylic acid methyl ester, methyl alpha-hydroxymethyl acrylate | |
COC(=O)C(=C)CO | |
116.12 | |
11815329 | |
85% |
Chemical Identifiers
15484-46-5 | |
116.12 | |
RFUCOAQWQVDBEU-UHFFFAOYSA-N | |
11815329 | |
COC(=O)C(=C)CO |
C5H8O3 | |
MFCD12545894 | |
methyl 2-hydroxymethyl acrylate, methyl 2-hydroxymethyl prop-2-enoate, methyl2-hydroxymethyl acrylate, methyl-2-hydroxymethyl-acrylate, pubchem17373, methyl 2-hydroxymethylacrylate, methyl alpha-hydroxymethylacrylate, 2-hydroxymethylacrylate methyl ester, 2-methylolacrylic acid methyl ester, methyl alpha-hydroxymethyl acrylateShow More | |
methyl 2-(hydroxymethyl)prop-2-enoate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
RUO â Research Use Only