missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(hydroxymethyl)acrylate, 85%, stabilized with 0.1 wt.% MEHQ, Thermo Scientific™

Chemical Identifiers

CAS 15484-46-5 , 150-19-6
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD12545894
InChI Key RFUCOAQWQVDBEU-UHFFFAOYSA-N
Synonym methyl 2-hydroxymethyl acrylate, methyl 2-hydroxymethyl prop-2-enoate, methyl2-hydroxymethyl acrylate, methyl-2-hydroxymethyl-acrylate, pubchem17373, methyl 2-hydroxymethylacrylate, methyl alpha-hydroxymethylacrylate, 2-hydroxymethylacrylate methyl ester, 2-methylolacrylic acid methyl ester, methyl alpha-hydroxymethyl acrylate
PubChem CID 11815329
IUPAC Name methyl 2-(hydroxymethyl)prop-2-enoate
SMILES COC(=O)C(=C)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC435900010
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC435900050
View Documents Promotion Details
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

15484-46-5 , 150-19-6
116.12
RFUCOAQWQVDBEU-UHFFFAOYSA-N
11815329
COC(=O)C(=C)CO
C5H8O3
MFCD12545894
methyl 2-hydroxymethyl acrylate, methyl 2-hydroxymethyl prop-2-enoate, methyl2-hydroxymethyl acrylate, methyl-2-hydroxymethyl-acrylate, pubchem17373, methyl 2-hydroxymethylacrylate, methyl alpha-hydroxymethylacrylate, 2-hydroxymethylacrylate methyl ester, 2-methylolacrylic acid methyl ester, methyl alpha-hydroxymethyl acrylate
methyl 2-(hydroxymethyl)prop-2-enoate

Specifications

15484-46-5 , 150-19-6
71°C to 72°C (2.0 Torr)
Authentic
Glass Bottle
MFCD12545894
methyl 2-hydroxymethyl acrylate, methyl 2-hydroxymethyl prop-2-enoate, methyl2-hydroxymethyl acrylate, methyl-2-hydroxymethyl-acrylate, pubchem17373, methyl 2-hydroxymethylacrylate, methyl alpha-hydroxymethylacrylate, 2-hydroxymethylacrylate methyl ester, 2-methylolacrylic acid methyl ester, methyl alpha-hydroxymethyl acrylate
COC(=O)C(=C)CO
116.12
11815329
85%
1.1290g/mL
90°C
84% min. (GC)
C5H8O3
1.129
RFUCOAQWQVDBEU-UHFFFAOYSA-N
methyl 2-(hydroxymethyl)prop-2-enoate
0.1 wt% Monomethylether hydroquinone
116.12
Methyl 2-(hydroxymethyl)acrylate, Stabilized
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only