missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Synthetic (Methyl Alcohol), Reagents
Methanol, Synthetic (Methyl Alcohol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2121000500C
Description
Greener Choice Claims
- Environmental benefits include:
- Source Reduction
Specifications
Methanol, Synthetic (Methyl Alcohol) | |
67-56-1 | |
64.5°C | |
Glass Bottle | |
0.7866 | |
Methyl Alcohol, Wood Alcohol | |
CO | |
32.04 |
-98°C | |
Colorless | |
500 mL | |
Liquid | |
CH4O | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
Reagent |
Chemical Identifiers
67-56-1 | |
32.04 | |
Methyl Alcohol, Wood Alcohol | |
CO |
CH4O | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol |