missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Synthetic (Methyl Alcohol), Reagents

Catalog No. p-200020924
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337524 1 L
01337523 200 L
01337522 20 L
01337521 4 L
01337520 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01337524 Supplier Labchem Inc Supplier No. C21210001C
Only null left

Methanol, Synthetic (Methyl Alcohol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl Alcohol, Wood Alcohol
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Boiling Point 64.5°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.7866
Grade Reagent
Chemical Name or Material Methanol, Synthetic (Methyl Alcohol)