missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol Anhydrous ACS, MilliporeSigma™
Supplier: Technidata MX04875
Specifications
Methanol | |
67-56-1,7732-18-5 | |
Glass Bottle | |
CH3OH | |
0.03% max | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 |
Anhydrous | |
4 L | |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
ACS Reagent |
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spiritShow More | |
CHEBI:17790 | |
CO |