missing translation for 'onlineSavingsMsg'
Learn More

Methanol Anhydrous ACS, MilliporeSigma™

Catalog No. p-3176677
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Glass Bottle
Metal Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMX04876 500 mL Glass Bottle
MMX04875 4 L Glass Bottle
MMX04873 20 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MMX04876 Supplier Technidata Supplier No. MX04876
Only null left

Chemical Identifiers

CAS 67-56-1 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Quantity 500 mL
Packaging Glass Bottle
Linear Formula CH3OH
Water 0.03% max
Formula Weight 32.04
Grade ACS
Chemical Name or Material Methanol, Anhydrous