Learn More
meso-Erythritol, 99%
CAS: 149-32-6 | C4H10O4 | 122.12 g/mol
Supplier: Thermo Scientific Chemicals 117821000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Renewable Energy
Specifications
meso-Erythritol | |
149-32-6 | |
White | |
Authentic | |
Plastic bottle | |
HOCH2[CH(OH)]2CH2OH | |
100 g | |
15,3733 | |
(10% in water) Clear colorless | |
OCC(O)C(O)CO | |
122.12 | |
CHEBI:17113 | |
99% |
99% | |
117.0°C to 121.0°C | |
329.0°C to 331.0°C | |
98.5% min. (ELSD) (HPLC) | |
C4H10O4 | |
MFCD00004710 | |
01,525 | |
erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythrite | |
UNXHWFMMPAWVPI-UHFFFAOYNA-N | |
(2S,3R)-butane-1,2,3,4-tetrol | |
222285 | |
122.12 | |
Crystalline Powder or Crystals |
Chemical Identifiers
149-32-6 | |
122.12 | |
UNXHWFMMPAWVPI-UHFFFAOYNA-N | |
222285 | |
OCC(O)C(O)CO |
C4H10O4 | |
MFCD00004710 | |
erythritol, meso-erythritol, phycitol, erythrit, phycite, erythrol, 2r,3s-butane-1,2,3,4-tetrol, erythro-tetritol, mesoerythritol, erythriteShow More | |
CHEBI:17113 |
Safety and Handling
EINECSNumber : 205-737-3
RTECSNumber : KF2000000
RUO – Research Use Only