missing translation for 'onlineSavingsMsg'
Learn More

Mesityl oxide, 99%, mixture of alpha- and beta-isomers

CAS: 141-79-7 | C6H10O | 98.14 g/mol

Supplier:  Thermo Scientific Chemicals 125600025

 View more versions of this product

Catalog No. AC125600025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 205-502-5

Recommended Storage : Product may darken during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only