missing translation for 'onlineSavingsMsg'
Learn More

Mesityl oxide, 99%, mixture of alpha- and beta-isomers

Catalog No. p-4509797
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125600010 1 L Glass Bottle
AC125600025 2.5 L Glass Bottle
AC125600100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC125600010 Supplier Thermo Scientific Chemicals Supplier No. 125600010
Only null left

CAS: 141-79-7 | C6H10O | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141-79-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
InChI Key SHOJXDKTYKFBRD-UHFFFAOYSA-N
Synonym mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone
PubChem CID 8858
IUPAC Name 4-methylpent-3-en-2-one
SMILES CC(=CC(=O)C)C

Specifications

Melting Point -53°C
Density 0.8540g/mL
Boiling Point 129°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 90% alpha-Isomer (GC)
Packaging Glass Bottle
Refractive Index 1.4420 to 1.446
Linear Formula (CH3)2C=CHCOCH3
Quantity 1 L
Beilstein 01,736
Merck Index 15,5978
Specific Gravity 0.854
Solubility Information Solubility in water: 28g/L (20°C). Other solubilities: miscible with most common organic solvents
Viscosity 0.64 mPa.s (20°C)
Formula Weight 98.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Mesityl oxide
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 205-502-5
Recommended Storage Product may darken during storage

RUO – Research Use Only