Learn More
Dual Manufacturing Company Mesh Series Sieves DFS Item
5 in. diameter (12.7 cm) sieves
Supplier:  Dual Manufacturing Company DULMG510
 View more versions of this product
Catalog No. 14-295A
Only null left
Explore available promotions
Description
Description
  • 10- to 80-mesh sieves made of brass; 100- to 200-mesh sieves of phosphor bronze
  • Gauze is spun into brass frames for tight, nonsagging sieve surfaces
  • Frames withstand rough use and fit together for stacking
  • 5 I.D. x 1.5 in.H (12.7 x 3.8cm)
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Specifications
1.5 in.
No. 10
Brass
Sieve
Brass
10
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories Promotions
Promotions