missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MERS Coronavirus Spike Protein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen PA581780
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for MERS-CoV (NCoV/Novel coronavirus) Spike.
MERS Coronavirus Spike Protein Polyclonal antibody specifically detects MERS Coronavirus Spike Protein in Virus samples. It is validated for ELISA.Specifications
MERS Coronavirus Spike Protein | |
Polyclonal | |
Unconjugated | |
MERS Coronavirus S protein; Spike glycoprotein | |
Recombinant MERS-CoV (NCoV/Novel coronavirus) Spike Protein (ECD, aa 1-1297) | |
100 ÎĽL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |