Learn More
Manganese(II) chloride tetrahydrate, 99.99% (metals basis)
CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol
Supplier: Thermo Scientific Chemicals 04444206
Description
It is used as a source of manganese in biological research, in the production of dry cell batteries, and precursor to the antiknock compound. It is used in industrial applications such as dyeing disinfecting, the purification of natural gas, and as a drying agent for linseed oil. It is used in pigment manufacturing, fertilizers and in construction materials and also used in aluminum smelting.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Manganese (II) chloride tetrahydrate | |
13446-34-9 | |
5 g | |
Cl2H8MnO4 | |
14,5728 | |
CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
manganese(2+) tetrahydrate dichloride | |
197.90 (125.84 Anhydrous) | |
Odorless | |
2.010 g/mL |
58°C | |
Crystalline | |
(metals basis) | |
MFCD00149792 | |
Soluble in water,acetic acid,pyridine,and ethanol but insoluble in ether. | |
O.O.O.O.[Cl-].[Cl-].[Mn++] | |
197.90 | |
99.99% | |
Hygroscopic |
Chemical Identifiers
13446-34-9 | |
197.90 | |
CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
O.O.O.O.[Cl-].[Cl-].[Mn++] |
Cl2H8MnO4 | |
MFCD00149792 | |
manganese(2+) tetrahydrate dichloride |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c
H302-H318-H373
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 231-869-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only