missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride tetrahydrate, 99.99% (metals basis)

Catalog No. p-7059272
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4444206 5 g
AA4444214 25 g
AA4444222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4444206 Supplier Thermo Scientific Chemicals Supplier No. 04444206
Only null left

CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol

It is used as a source of manganese in biological research, in the production of dry cell batteries, and precursor to the antiknock compound. It is used in industrial applications such as dyeing disinfecting, the purification of natural gas, and as a drying agent for linseed oil. It is used in pigment manufacturing, fertilizers and in construction materials and also used in aluminum smelting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58°C
Physical Form Crystalline
Quantity 5 g
Merck Index 14,5728
Solubility Information Soluble in water,acetic acid,pyridine,and ethanol but insoluble in ether.
Formula Weight 197.90 (125.84 Anhydrous)
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.010 g/mL
Assay (metals basis)
Chemical Name or Material Manganese (II) chloride tetrahydrate
Show More Show Less
Hazard Category H302-H318-H373
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-869-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only