missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
GREENER_CHOICE

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$256.71 - $513.41

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PLMG2-2Y
SDS View Documents
SPEX CertiPrep Group LLC
PLMG22Y
125 mL Triple-leached LDPE bottle
Each for $256.71
Only null left
 
PLMG2-2X
SDS View Documents
SPEX CertiPrep Group LLC
PLMG22X
500 mL Triple-leached LDPE bottle
Each for $513.41
Only null left
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Mg1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
missing translation for 'documents'

missing translation for 'documents'

Product intended for laboratory use only