missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit

Catalog No. A48383R
Encompass
Change view
Click to view available options
Quantity:
2000 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A48383R 2000 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A48383R Supplier Applied Biosystems™ Supplier No. A48383R

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The MagMAX Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit recovers RNA and DNA from virus and gram-negative bacteria in samples such as blood, swabs, urine, and transport media.

  • Automation-ready protocols designed for the KingFisher Flex and KingFisher Duo Prime systems
  • Fast procedure allows for 96 samples to be processed in <30 minutes
  • Flexible protocol accommodates sample volume inputs from 200 μL to 2 mL of biofluids or transfer media
  • No need for carrier RNA
  • Elution volumes ranging from 50 to 100 μL
  • The optimized reagents included allow you to maximize the amount of sample input
  • Up to 400 μL of sample can be processed per well in a KingFisher 96-deep well plate.
  • Protocols are available to process up to 2 mL of sample per well in a KingFisher 24-deep well plate

Specifications

Content And Storage • 550 mL Binding Solution, store at room temperature
• 1000 mL Wash Solution, store at room temperature
• 100 mL Elution Solution, store at room temperature
• 10 mL Proteinase K, store at room temperature
• 20 mL DNA/RNA Binding Beads, store at room temperature

Format 96 Deep-well Plate, 24 Deep-well Plate
Purification Time 30 min.
No. of Reactions 2000 Preps
Product Line MagMAX
Quantity 2000 preps
Shipping Condition Room Temperature
Do you offer the Proteinase K that is included in the MagMAX Viral/Pathogen II (MVP II) Nucleic Acid Isolation Kit (Cat. No. A48383) as a stand-alone product?

No standalone components are validated for In Vitro Diagnostic (IVD) use with this kit.

For Research Use Only. Not for use in diagnostic procedures.