missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tert-butoxide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4413309

 View more versions of this product

Catalog No. AA4413309



Specifications

Specifications

Lithium tert-butoxide
Air and moisture sensitive
~150°C (decomposition)
1907-33-1
C4H9LiO
MFCD00050479
3620018
Soluble in toluene,hexane,tetrahydrofuran and methyl tert-butyl ether.
[Li+].CC(C)(C)[O-]
80.055
80.06
99.9%
Odorless
White to Cream
10g
(metals basis)
(CH3)3COLi
UN3206
2-methyl-2-propanol lithium salt
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium;2-methylpropan-2-olate
23664764
Powder

Chemical Identifiers

1907-33-1
80.055
LZWQNOHZMQIFBX-UHFFFAOYSA-N
23664764
[Li+].CC(C)(C)[O-]
C4H9LiO
MFCD00050479
2-methyl-2-propanol lithium salt
lithium;2-methylpropan-2-olate
Safety and Handling

Safety and Handling

GHS H Statement
H251-H314-H302
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Harmful if swallowed.

P210-P240-P241-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H228-H302-H314

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 217-611-5

RTECSNumber : UB8520000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents