missing translation for 'onlineSavingsMsg'
Learn More

Lithium tert-butoxide, 99.9% (metals basis)

Catalog No. p-7059173
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4413309 10 g
AA4413318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4413309 Supplier Thermo Scientific Chemicals Supplier No. 04413309
Only null left

CAS: 1907-33-1 | C4H9LiO | 80.055 g/mol

Lithium tert-butoxide is a mild base used in organic synthesis. It is involved in the alfa-alkylation reaction of ketones with primary alcohols. It is utilized as a catalyst in the reduction of ketones to the corresponding secondary alcohols. It acts as lithium source utilized in the preparation of LixTiyOz thin films. Further, it is used in conjunction with organometallic reagents/catalysts to modify their reactivity and selectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1907-33-1
Molecular Formula C4H9LiO
Molecular Weight (g/mol) 80.055
MDL Number MFCD00050479
InChI Key LZWQNOHZMQIFBX-UHFFFAOYSA-N
Synonym lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate
PubChem CID 23664764
IUPAC Name lithium;2-methylpropan-2-olate
SMILES [Li+].CC(C)(C)[O-]

Specifications

Melting Point ∼150°C (decomposition)
Color White to Cream
Odor Odorless
Linear Formula (CH3)3COLi
Quantity 10 g
UN Number UN3206
Beilstein 3620018
Sensitivity Air and moisture sensitive
Solubility Information Soluble in toluene,hexane,tetrahydrofuran and methyl tert-butyl ether.
Formula Weight 80.06
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Lithium tert-butoxide
Show More Show Less
Hazard Category H228-H302-H314
Hazard Statement GHS H Statement
H251-H314-H302
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement P210-P240-P241-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 217-611-5
RTECSNumber UB8520000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only