Learn More
Lithium nitrate, anhydrous, 99.999% (metals basis)
CAS: 7790-69-4 | LiNO3 | 68.94 g/mol
Supplier: Thermo Scientific Chemicals 04445618
Description
Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent used in the production of red-colored fireworks and flares, as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from sun for cooking. It is applied to concrete-pavement to withstand weathering effects.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
600°C (decomposition) | |
264°C | |
7790-69-4 | |
Crystalline/Lump | |
(metals basis) | |
MFCD00011094 | |
14,5536 | |
Soluble in water,ethanol,methanol,pyridine,ammonia and acetone. | |
[Li+].[O-][N+]([O-])=O | |
68.94 | |
CHEBI:63315 | |
99.999% | |
2.38 g/mL |
Lithium nitrate | |
Anhydrous | |
White | |
50 g | |
LiNO3 | |
UN2722 | |
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure | |
IIPYXGDZVMZOAP-UHFFFAOYSA-N | |
lithium;nitrate | |
10129889 | |
68.95 | |
Odorless |
Chemical Identifiers
7790-69-4 | |
68.94 | |
IIPYXGDZVMZOAP-UHFFFAOYSA-N | |
10129889 | |
lithium;nitrate |
LiNO3 | |
MFCD00011094 | |
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure | |
CHEBI:63315 | |
[Li+].[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
H272-H302-H319
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: LITHIUM NITRATE
EINECSNumber : 232-218-9
RTECSNumber : QU9330000
TSCA : Yes
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only