missing translation for 'onlineSavingsMsg'
Learn More

Lithium nitrate, anhydrous, 99.999% (metals basis)

Catalog No. p-7059352
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4445609 10 g
AA4445618 50 g
AA4445630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4445609 Supplier Thermo Scientific Chemicals Supplier No. 04445609
Only null left

CAS: 7790-69-4 | LiNO3 | 68.94 g/mol

Lithium nitrate is used in ceramics, pyrotechnics, salt baths, heat-exchange media, refrigeration systems, rocket propellant and in solar batteries. It acts as an oxidizing agent used in the production of red-colored fireworks and flares, as a catalyst to accelerate the breakdown of nitrogen oxides and as a medium to store heat received from sun for cooking. It is applied to concrete-pavement to withstand weathering effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
Synonym lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
PubChem CID 10129889
ChEBI CHEBI:63315
IUPAC Name lithium;nitrate
SMILES [Li+].[O-][N+]([O-])=O

Specifications

Boiling Point 600°C (decomposition)
Melting Point 264°C
Color White
Physical Form Crystalline/Lump
Quantity 10 g
UN Number UN2722
Merck Index 14,5536
Solubility Information Soluble in water,ethanol,methanol,pyridine,ammonia and acetone.
Formula Weight 68.95
Percent Purity 99.999%
Odor Odorless
Density 2.38 g/mL
Assay (metals basis)
Chemical Name or Material Lithium nitrate, Anhydrous
Show More Show Less
Hazard Category H272-H302-H319
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: LITHIUM NITRATE
EINECSNumber 232-218-9
RTECSNumber QU9330000
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only