Learn More
Lithium hydroxide, anhydrous, 99.995% (metals basis)
CAS: 1310-65-2 | HLiO | 23.95 g/mol
Supplier: Thermo Scientific Chemicals 04447114
Description
It is used as a heat transfer medium, as a storage-battery electrolyte and also used for the production of lithium greases. It is also used in ceramics, in some portland cement formulations, in the absoption of carbondioxide from sealed enviornments such as submarines, spacecrafts and breathing apparatus. It is used in esterification reactions, as stabilizer in photographic developments and as a coolant in pressurized water reactors for corrosion control.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Environmental benefits include:
- Recycled Content
- Free-of a substance that causes environmental harm
Specifications
924°C | |
462°C | |
1310-65-2 | |
14 | |
25 g | |
HLiO | |
UN2680 | |
lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous | |
WMFOQBRAJBCJND-UHFFFAOYSA-M | |
lithium(1+) hydroxide | |
3939 | |
23.95 | |
Odorless | |
2.54 g/mL |
Lithium hydroxide | |
Anhydrous | |
White | |
Powder | |
(metals basis) | |
MFCD00011095 | |
14,5534 | |
Soluble in water. Slightly soluble in ethanol and methanol | |
[Li+].[OH-] | |
23.95 | |
CHEBI:33979 | |
99.995% | |
Air sensitive; Hygroscopic |
Chemical Identifiers
1310-65-2 | |
23.95 | |
WMFOQBRAJBCJND-UHFFFAOYSA-M | |
3939 | |
lithium(1+) hydroxide |
HLiO | |
MFCD00011095 | |
lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrousShow More | |
CHEBI:33979 | |
[Li+].[OH-] |
Safety and Handling
GHS H Statement
H301-H331-H314-H318
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335-H336-H373
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: LITHIUM HYDROXIDE
EINECSNumber : 215-183-4
RTECSNumber : OJ6307070
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only