missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydroxide, anhydrous, 99.995% (metals basis)

Catalog No. p-7059319
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AA4447106 5 g
AA4447114 25 g
AA4447122 100 g
3 options

Catalog No. AA4447106

Supplier: Thermo Scientific Chemicals 04447106

Only null left

CAS: 1310-65-2 | HLiO | 23.95 g/mol

It is used as a heat transfer medium, as a storage-battery electrolyte and also used for the production of lithium greases. It is also used in ceramics, in some portland cement formulations, in the absoption of carbondioxide from sealed enviornments such as submarines, spacecrafts and breathing apparatus. It is used in esterification reactions, as stabilizer in photographic developments and as a coolant in pressurized water reactors for corrosion control.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1310-65-2
Molecular Formula HLiO
Molecular Weight (g/mol) 23.95
MDL Number MFCD00011095
InChI Key WMFOQBRAJBCJND-UHFFFAOYSA-M
Synonym lithium hydroxide, lithium hydrate, lioh, lithium hydroxide anhydrous, lithium hydroxide li oh, lithiumhydroxid, lithiumhydroxide, lithium hydoxide, unii-903yl31jas, lithium hydroxide, anhydrous
PubChem CID 3939
ChEBI CHEBI:33979
IUPAC Name lithium(1+) hydroxide
SMILES [Li+].[OH-]

Specifications

Boiling Point 924°C
Melting Point 462°C
Color White
pH 14
Physical Form Powder
Quantity 5 g
UN Number UN2680
Merck Index 14,5534
Solubility Information Soluble in water. Slightly soluble in ethanol and methanol
Formula Weight 23.95
Percent Purity 99.995%
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 2.54 g/mL
Assay (metals basis)
Chemical Name or Material Lithium hydroxide, Anhydrous
Show More Show Less
Hazard Category H302-H314-H335-H336-H373
Hazard Statement GHS H Statement
H301-H331-H314-H318
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: LITHIUM HYDROXIDE
EINECSNumber 215-183-4
RTECSNumber OJ6307070
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only