Learn More
Labconco™ Purifier™ HEPA-Filtered Safety Enclosure, Without Guardian Monitor
Ensure personnel and environmental biohazard protection by outfitting your lab with the Labconco Purifier™ HEPA-Filtered Safety Enclosure.
$11355.95 - $15082.95
Specifications
Product Type | HEPA Filtered Enclosure |
---|---|
Height (English) | 37.08 in. |
Sash Opening (Metric) | 23.97 cm |
Filter Type | HEPA |
Housing Material | Epoxy Coated Aluminum and Steel |
Catalog Number | Mfr. No. | Product Type | Electrical Requirements | Width (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Electrical Requirements | Width (Metric) | Price | Quantity | |||
10-304-126
|
Labconco™
3980300 |
HEPA Filtered Enclosure | 115 V, 60 Hz | 90.8 cm |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
10-304-130
|
Labconco™
3980400 |
HEPA Filtered Enclosure | 115 V, 60 Hz | 121.28 cm |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
10-304-122
|
Labconco™
3980200 |
HEPA Filtered Enclosure | 115 V, 60 Hz | 60.32 cm |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Meet requirements for Biosafety Level 1, 2, or 3 containment without product protection, making these an economical alternative to Class II biosafety cabinets. Also suitable for chemical and particulate powder weighing and other procedures generating fine dusts or aerosols. HEPA-filtered enclosures require no outside exhausting.
- Total exhaust from 155 to 310cfm, depending on hood width
- Sash pivots 10°, from 9.5 in. (24cm) operating position to 20 in. (50.8cm) opening for loading and maintenance
- Quiet variable-speed, solid-state internal blower maintains appropriate air velocity
- Airfoil with Clean-Sweep™ airflow openings
- Front-mounted pressure gauge monitors pressure differential against the HEPA filter
- HEPA filter is 99.99% efficient on 0.3ÎĽm particles (where applicable)
- Negative pressure design ensures containment—should a leak occur, the system traps the contaminated air and recirculates it through the filter
- 0.25 in. (6.4mm) Tempered glass side panels and sash
- Glare-free fluorescent lamp mounted outside contamination area
- Blower and light switches are front-mounted for easy access
- Powder-coated aluminum frame and steel rear plenum and zone-perforated rear baffle
- Back panel has two utility ports for service connections
- Open bottom allows enclosure to be placed on existing cabinetry or accessory dished work surface
Order Info
28.75L x 37 in. H (73 x 94cm). 115V 60Hz, 3A. Models with Guardian Monitor are also available (Cat. No. 10-304-125, -129, -133, -127 and -131).
Certifications
ETL listed (certified to UL 3101-1/61010-1 and CAN/CSA C22.2 No. 1010.1).
Compliance
ASHRAE™ 110-1995 conformity tested. Biological containment verified per NSF Standard Number 49 Personnel Protection Test.
Specifications
HEPA Filtered Enclosure | |
23.97 cm | |
Epoxy Coated Aluminum and Steel | |
94.18 cm |
37.08 in. | |
HEPA | |
Fluorescent |