Learn More
DWK Life Sciences™ Kimble™ Kontes™ Bubbler Tube, Monier-Williams, 24/40 Joint
Component Parts of Monier-Williams Sulfites Distillation Apparatus which is used for the determination of free sulfite and a reproducible portion of the bound sulfites in food, such as the carbonyl addition products.
Supplier: DWK Life Sciences™ 5138020000
Description
- Bubbler Tube
- Standard Taper joint 24/40
- Case of 1
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Bubbler Tube | |
24/40 |
Promotions
-
DWK Life Sciences
,
DWK Life Sciences (Wheaton)
Get $500 or $2,000 in DWK Products with Purchase
Spend $500 or $2,000 on DWK Life Sciences products, get an equal amount to buy products at no additional cost.Expires: 06/30/25 Redemption Only
Limited Time See Full Promo Details