DWK Life Sciences Kimble™ Kontes™ Class A Volumetric Flasks with Std. Taper Stopper
Calibrated “To Contain”
Manufacturer: DWK Life Sciences KC280142000
Description
- KimCote™ plastic coating for added safety
- With single graduation ring blasted on the neck
- standard taper ground glass stopper is supplied
- Marking spots are durable white ceramic enamel
- Designed from ASTM™ Specification E288, Class A unserialized requirements
A world of expertise, in your hands. Visit www.fishersci.com to learn more.
Specifications
27 | |
2000 mL | |
15.74 in. | |
±0.5 mL |
Glass | |
Borosilicate Glass | |
40 cm |