Learn More
DWK Life Sciences Kimble™ Kontes™ Class A Volumetric Flasks with Std. Taper Stopper
Calibrated To Contain
$1101.30 - $4313.87
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
01183005
|
DWK Life Sciences
KC28014250 |
250 mL |
Case of 12 Each for $3,482.40
|
|
|||||
|
12-141-348
|
DWK Life Sciences
KC28014500 |
500 mL |
Case of 12 Each for $4,313.87
|
|
|||||
|
12-141-346
|
DWK Life Sciences
KC28014100 |
100 mL |
Case of 12 Each for $2,786.90
|
|
|||||
|
12-141-349
|
DWK Life Sciences
KC280141000 |
1000 mL |
Case of 4 Each for $2,011.00
|
|
|||||
|
12-141-350
|
DWK Life Sciences
KC280142000 |
2000 mL |
Case of 2 Each for $1,101.30
|
|
|||||
Description
- KimCote™ plastic coating for added safety
- With single graduation ring blasted on the neck
- standard taper ground glass stopper is supplied
- Marking spots are durable white ceramic enamel
- Designed from ASTM™ Specification E288, Class A unserialized requirements
A world of expertise, in your hands. Visit www.fishersci.com to learn more.
Specifications
| Glass |