missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Flip, Tear-Off Button Top Aluminum Seals, Color Coded

Designed to secure septa and stoppers to bottles and vials
$1311.97 - $1311.97
Specifications
Material | Aluminum |
---|---|
Autoclavable | Autoclavable |
Quantity | 1000 Cs. |
Type | Flip-up Button-top Cap |
Closure Size | 13 mm |
Catalog Number | Mfr. No. | Color | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | ||||||
03-340-333
|
DWK Life Sciences
73844B13 |
Blue |
Case of 1000 Each for $1,311.97
|
|
||||||
03-340-334
|
DWK Life Sciences
73844C13 |
Red |
Case of 1000 Each for $1,311.97
|
|
||||||
Description
- Tamper-evident flip, tear-off button top protects target area and allows easy removal of the aluminum seal
- For use with 13mm serum finished bottles and vials
- Available with blue, red, and white plastic buttons to help separate and identify samples
Specifications
Aluminum | |
1000 Cs. | |
13 mm |
Autoclavable | |
Flip-up Button-top Cap |