missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech™ Pure G5 Sterile Latex Gloves
GREENER_CHOICE

Sterile, hand-specific latex gloves provide sensitive and dextrous protection in Class 100/ISO 5 or higher cleanroom environments.

$540.93 - $540.93

Specifications

Certifications/Compliance ASTM D3577, IEST-RP-CC005
Certifications/Compliance (Europe) PPE Cat III, EN ISO 374-1:2016, EN 374-4:2003, EN ISO 374-5:2016
Cleanroom Class Class 100 (ISO 5)
AQL 1.5
Color Natural
View More Specs

Alert:

This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-048586H
View Documents
Ansell
HC1110S
10
Case of 200 Pair for $540.93
Only null left
 
Description

Description

  • Fully sterilized latex material provides high levels of contamination protection
  • Hand-specific, disposable and a natural color
  • Textured palm and fingertips enhance grip and tactile sensitivity
  • Beaded cuffs add strength to the gloves reducing the risk of tearing
  • Easy donning and doffing
  • Made from latex sterilized with gamma radiation and containing no powder
  • Suitable for use in EU GMP ISO Class 5 Grade A or higher sterile cleanrooms
Specifications

Specifications

ASTM D3577, IEST-RP-CC005
Class 100 (ISO 5)
Natural
Fully Textured
Latex
Sterile
0.20 mm
12 in.
PPE Cat III, EN ISO 374-1:2016, EN 374-4:2003, EN ISO 374-5:2016
1.5
Single-use
Specific
Powder Free
8 mil
Kimtech™ Pure G5
30.5 cm
Videos
SDS
Documents

Documents