missing translation for 'onlineSavingsMsg'
Learn More

Justriteā„¢ Standard Aerosolvā„¢ Aerosol Can Disposal System: Cartridges and Filters
GREENER_CHOICE

For use with Serosolv Can Disposal System

$377.58 - $487.85

Specifications

Material Carbon
View More Specs

Products 3
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
19-077307
View Documents
Justriteā„¢
28198
Activated Carbon Cartridge N/A
Only null left
 
19077306
View Documents
Justriteā„¢
28197
Combination Coalescing/Carbon Filter N/A
Only null left
 
29-635-8037
View Documents
Justriteā„¢
28224
Color-Changing Combination Coalescing/Carbon Filter N/A
Only null left
 
Description

Description

Replaxement cartridges and filters.

Specifications

Specifications

Carbon
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'