missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Standard Aerosolv™ Aerosol Can Disposal System: Cartridges and Filters

For use with Serosolv Can Disposal System

Supplier:  Justrite™ 28224

 View more versions of this product

Catalog No. 296358037


Only null left

Description

Description

Replacement cartridges and filters.

Specifications

Chemical Identifiers

Carbon

Specifications

EPA, 40CRF261.23(a)(6)
For aerosolv aerosol can disposal system
Carbon
Color-Changing Combination Coalescing/Carbon Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions