Learn More
Mystaire™ Isola™ Filtered Workstation Dual Purpose Safety Filters
For use with Isola™ filtered workstations. Provides operator safety from potentially harmful fumes, odors and airborne particulate.
Supplier:  Mystaire™ MY6500
 View more versions of this product
Catalog No. 15-338-953
Only null left
Explore available promotions
Description
Description
  • Designed to incorporate both gas phase and mechanical phase filtration into one safety filter
  • Provides adequate reserve gas phase capacity as the primary filter(s) reach saturation
Specifications
Specifications
Particulate
2 in.
12 in.
51 mm
305 mm
Dual Purpose Safety Filter
Isola Filtered Workstation (48 in.)
15 lb.
20 in.
305 x 508 x 51 mm (12 x 20 x 2 in.)
508 mm
Bonded Carbon, HEPA
Product Suggestions
Product Suggestions