Learn More
Iron boride, 98%
Used to improve abrasion resistance, corrosion resistance, wear resistance and oxidation resistance. It is also used in oil and gas refinery, chemical extraction, agricultural and textile extrusion. | CAS: 12006-84-7 | BFe | 66.66 g/mol
Supplier: Thermo Scientific Chemicals 08814614
Description
Iron boride is used to improve abrasion resistance, corrosion resistance, wear resistance and oxidation resistance. It is also used in oil and gas refinery, chemical extraction, agricultural and textile extrusion. Being inexpensive as well as having excellent mechanical, frictional and corrosion resistant properties, it can be used as replacement of ceramic or cermet type of materials in fabrication and machining. It is also involved in the production of borated steels and borated stainless steels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Iron boride | |
Powder | |
BFe | |
iron boride, iron boride feb, ferrous boride, ironylidyneborane, ferrioborane, ferric boride | |
UJYBTTFGLAOKMN-UHFFFAOYSA-N | |
boranylidyneiron | |
82789 | |
98% | |
7.15 |
12006-84-7 | |
25 g | |
MFCD00016092 | |
Insoluble in water. | |
[B+]=[Fe-] | |
66.66 | |
66.66 | |
Glass bottle | |
7.15 |
Chemical Identifiers
12006-84-7 | |
66.66 | |
UJYBTTFGLAOKMN-UHFFFAOYSA-N | |
82789 |
BFe | |
MFCD00016092 | |
iron boride, iron boride feb, ferrous boride, ironylidyneborane, ferrioborane, ferric borideShow More | |
[B+]=[Fe-] |
Safety and Handling
EINECSNumber : 234-489-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only